Merck
  • Up-Regulation of KPNB1 Involves in Neuronal Apoptosis Following Intracerebral Hemorrhage in Adult Rats.
Up-Regulation of KPNB1 Involves in Neuronal Apoptosis Following Intracerebral Hemorrhage in Adult Rats.
Neurochemical research (2015-08-26)
Aihua Dai, Xiaorong Liu, Yu Zhang, Lijian Han, Liang Zhu, Haidan Ni, Rongrong Chen, Maohong Cao
ABSTRACT
Kpnb1, also known as Importin β1, is a member of the Karyopherin protein family which plays a important role in nuclear import and export pathways. Its expression has been shown to be responsive to stress, such as heat shock, ethanol and oxidative stress. Previous studies demonstrated that Kpnb1 had anti-apoptotic in cervical cancer. These together prompted us to explore whether Kpnb1 has some association with neuron apoptosis in the pathophysiology of intracerebral hemorrhage (ICH). In our study, an ICH model was established by injecting into the right basal ganglia of adult rats with their autologous whole blood and assessed by behavioral tests. We found Kpnb1 were significantly up-regulated adjacent to the hematoma following ICH by Western blot and immunohistochemistry. Double immunofluorenscence manifested Kpnb1 was strikingly increased in neurons, not astrocytes or microglia. Furthermore, we also found that kpnb1 had co-localizations with active-caspase-3 which is a neuronal apoptosis marker suggesting its role in neuronal apoptosis. What's more, our in vitro study, using Kpnb1 RNA interference in PC12 cells, further indicated that Kpnb1 might exert its pro-apoptotic function on neuronal apoptosis. Therefore, Kpnb1 may play a role in the neuronal apoptosis following ICH.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human KPNB1
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kpnb1
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%