Merck
  • Endogenous myeloperoxidase is a mediator of joint inflammation and damage in experimental arthritis.
Endogenous myeloperoxidase is a mediator of joint inflammation and damage in experimental arthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2014-04-24)
Dragana Odobasic, Yuan Yang, Ruth C M Muljadi, Kim M O'Sullivan, Wenping Kao, Malcolm Smith, Eric F Morand, Stephen R Holdsworth
ABSTRACT
Myeloperoxidase (MPO) is implicated as a local mediator of tissue damage when released extracellularly in many chronic inflammatory diseases. The purpose of this study was to explore the role of endogenous MPO in experimental rheumatoid arthritis (RA). K/BxN serum-transfer arthritis was induced in C57BL/6 wild-type (WT) and MPO knockout (MPO(-/-) ) mice, and disease development was assessed. MPO activity was measured in joint tissues from mice with or without K/BxN arthritis. Collagen-induced arthritis (CIA) was induced in WT and MPO(-/-) mice, and disease development and immune responses were examined. MPO expression was assessed in synovial biopsy samples from patients with active RA, and the effect of MPO on synovial fibroblasts was tested in vitro. MPO was up-regulated in the joints of mice with K/BxN arthritis, and MPO deficiency attenuated the severity of the disease without affecting circulating cytokine levels. In CIA, MPO(-/-) mice had enhanced CD4+ T cell responses and reduced frequency of regulatory T cells in the lymph nodes and spleen, as well as augmented interleukin-17A and diminished interferon-γ secretion by collagen-stimulated splenocytes, without an effect on circulating anticollagen antibody levels. Despite enhanced adaptive immunity in secondary lymphoid organs, CIA development was attenuated in MPO(-/-) mice. Intracellular and extracellular MPO was detected in the synovium of patients with active RA, and human MPO enhanced the proliferation and decreased the apoptosis of synovial fibroblasts in vitro. MPO contributes to the development of arthritis despite suppressing adaptive immunity in secondary lymphoid organs. This suggests distinct effects of local MPO on arthritogenic effector responses.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard