Merck
  • Construction of dentate bonded TiO2-CdSe heterostructures with enhanced photoelectrochemical properties: versatile labels toward photoelectrochemical and electrochemical sensing.
Construction of dentate bonded TiO2-CdSe heterostructures with enhanced photoelectrochemical properties: versatile labels toward photoelectrochemical and electrochemical sensing.
Dalton transactions (Cambridge, England : 2003) (2014-11-20)
Picheng Gao, Hongmin Ma, Tao Yan, Dan Wu, Xiang Ren, Jiaojiao Yang, Bin Du, Qin Wei
ABSTRACT
A facile synthetic route for TiO2-CdSe heterostructures was proposed based on dentate binding of TiO2 to carboxyl. Carboxyl functionalized CdSe quantum dots (CF-CdSe QDs) were successfully bonded onto TiO2 nanoparticles (NPs), which could significantly improve the photoelectrochemical (PEC) properties of TiO2 NPs. This is ascribed to the fact that CdSe QDs with a narrow band gap could be stimulated under visible light irradiation, and the energy levels of TiO2 NPs and CF-CdSe QDs are aligned with an electrolyte solution. High resolution transmission electron microscopy images revealed the heterostructures of the TiO2-CdSe composites. Ultraviolet visible spectroscopy, photoluminescence emission spectroscopy and electrochemical impedance spectroscopy analysis exhibited that the prepared TiO2-CdSe heterostructures have improved light absorption, charge separation efficiency and electron transfer ability in the visible light region. TiO2-CdSe heterostructures were used as versatile labels for fabrication of PEC and electrochemical immunosensors, and human immune globulin G (HIgG) was used as a model analyte. The immunosensor showed high sensitivity, a low detection limit and a wide linear range, which could be applied in practical serum sample analysis. The constructed TiO2-CdSe heterostructures would have potential applications in photocatalysis, aptasensors, cytosensors and other areas of nanotechnology.
Sigma-Aldrich
Sodium sulfite, JIS special grade, ≥97.0%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) butoxide, purum, ≥97.0% (gravimetric)
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Sodium sulfite, analytical standard
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Sodium sulfite, ≥98%
Sigma-Aldrich
Sodium sulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Sigma-Aldrich
Sodium sulfite, puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Sigma-Aldrich
Sodium sulfite, ACS reagent, ≥98.0%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
1,2-Dichloroethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, JIS special grade, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%