Merck
  • Mitochondrial SIRT5 is present in follicular cells and is altered by reduced ovarian reserve and advanced maternal age.
Mitochondrial SIRT5 is present in follicular cells and is altered by reduced ovarian reserve and advanced maternal age.
Reproduction, fertility, and development (2013-08-28)
Leanne Pacella-Ince, Deirdre L Zander-Fox, Michelle Lane
ABSTRACT
Women with reduced ovarian reserve or advanced maternal age have an altered metabolic follicular microenvironment. As sirtuin 5 (SIRT5) senses cellular metabolic state and post-translationally alters protein function, its activity may directly impact on oocyte viability and pregnancy outcome. Therefore, we investigated the role of SIRT5 in relation to ovarian reserve and maternal age. Women (n=47) undergoing routine IVF treatment were recruited and allocated to one of three cohorts based on ovarian reserve and maternal age. Surplus follicular fluid, granulosa and cumulus cells were collected. SIRT5 mRNA, protein and protein activity was confirmed in granulosa and cumulus cells via qPCR, immunohistochemistry, western blotting and desuccinylation activity. The presence of carbamoyl phosphate synthase I (CPS1), a target of SIRT5, was investigated by immunohistochemistry and follicular-fluid ammonium concentrations determined via microfluorometry. Women with reduced ovarian reserve or advanced maternal age had decreased SIRT5 mRNA, protein and desuccinylation activity in granulosa and cumulus cells resulting in an accumulation of follicular-fluid ammonium, presumably via alterations in activity of a SIRT5 target, CPS1, which was present in granulosa and cumulus cells. This suggests a role for SIRT5 in influencing oocyte quality and IVF outcomes.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sirtuin 5 human, recombinant, expressed in E. coli, ≥50% (SDS-PAGE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard