Merck
  • Revised guidelines for the diagnosis and management of hairy cell leukaemia and hairy cell leukaemia variant*.
Revised guidelines for the diagnosis and management of hairy cell leukaemia and hairy cell leukaemia variant*.
British journal of haematology (2011-11-25)
Gail Jones, Nilima Parry-Jones, Bridget Wilkins, Monica Else, Daniel Catovsky
ABSTRACT
The British Committee for Standards in Haematology first produced guidelines for the diagnosis and management of hairy cell leukaemia and hairy cell leukaemia variant in 2000. This revision updates those guidelines and covers the areas of diagnosis, treatment and assessment of response to therapy.
Sigma-Aldrich
CD11c (EP157) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
CD11c (5D11) Mouse Monoclonal Antibody