Merck
  • Octaproline, a conformationally flexible chiral selector in liquid chromatographic enantioseparation.
Octaproline, a conformationally flexible chiral selector in liquid chromatographic enantioseparation.
Journal of chromatography. A (2014-05-28)
Arnau Novell, Cristina Minguillón
ABSTRACT
A proline octapeptide-derived chiral selector (CS) end-capped using a pivaloyl group was covalently linked to a silica gel chromatographic matrix by the C-terminal group. The chromatographic behaviour of the resulting chiral stationary phase (CSP) using different conditions was compared to those containing 3,5-dimethylphenylcarbamate residues on the proline units. An enantioseparation ability highly dependent on the mobile phase used is observed for these CSPs. When mixtures of alkane/alcohol or alkane/ether are used as mobile phase a similar enantioselectivity is obtained. Nevertheless, in the presence of chlorinated solvents, and without a hydrogen bonding donor in the mobile phase, enantioselectivity is extremely reduced. The reversibility of this phenomenon, attributed to a conformational change in the CS, is examined.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O