Merck
  • Factor XI regulates pathological thrombus formation on acutely ruptured atherosclerotic plaques.
Factor XI regulates pathological thrombus formation on acutely ruptured atherosclerotic plaques.
Arteriosclerosis, thrombosis, and vascular biology (2014-06-21)
Maurits L van Montfoort, Marijke J E Kuijpers, Véronique L Knaup, Sanjay Bhanot, Brett P Monia, Joris J T H Roelofs, Johan W M Heemskerk, Joost C M Meijers
ABSTRACT
Coagulation factor XI is proposed as therapeutic target for anticoagulation. However, it is still unclear whether the antithrombotic properties of factor XI inhibitors influence atherosclerotic disease and atherothrombosis. Our aim is to investigate whether factor XI antisense oligonucleotides could prevent thrombus formation on acutely ruptured atherosclerotic plaques. Atherosclerotic plaques in the carotid arteries of Apoe(-/-) mice were acutely ruptured using ultrasound. The subsequent thrombus formation was visualized and quantified by intravital microscopy and immunohistochemistry. Mice were pretreated with either factor XI antisense or nonsense oligonucleotides (50 mg/kg) to lower factor XI plasma levels. A tail bleeding assay was used to determine the safety. On plaque rupture, initial platelet adhesion and platelet plug formation were not impaired in animals treated with factor XI antisense oligonucleotides. However, the ensuing thrombus formation and fibrin deposition were significantly lower after 5 to 10 minutes (P<0.05) in factor XI antisense oligonucleotide-treated animals without inducing a bleeding tendency. Furthermore, thrombi from antisense-treated animals were less stable than thrombi from placebo-treated animals. Moreover, macrophage infiltration and collagen deposition were lower in the carotid arteries of factor XI antisense-treated animals. No neutrophils were present. Factor XI antisense oligonucleotides safely prevent thrombus formation on acutely ruptured atherosclerotic plaques in mice. Furthermore, perturbed carotid arteries from factor XI antisense-treated animals show a less severe inflammatory response.
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Fibrin from human plasma, insoluble powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance