Merck
  • Review article: delivery and efficacy of topical 5-aminosalicylic acid (mesalazine) therapy in the treatment of ulcerative colitis.
Review article: delivery and efficacy of topical 5-aminosalicylic acid (mesalazine) therapy in the treatment of ulcerative colitis.
Alimentary pharmacology & therapeutics (2011-03-10)
M S Harris, G R Lichtenstein
ABSTRACT
The use of topical therapy in the treatment of ulcerative colitis has declined in recent years despite evidence of good efficacy. To review US prescription trends for 5-aminosalicylic acid (5-ASA) since the US approval of Asacol extended-release oral mesalazine (mesalamine) in 1992; to estimate the optimal level of 5-ASA exposure in the distal colon; to determine factors influencing distal colonic exposures; and to compare the effectiveness of different 5-ASA formulations (oral, topical suspension, foam, suppositories) in clinical trials. Review of clinical trials, physiologic studies and prescription trends of various mesalazine formulations for treatment of distal ulcerative colitis. Between 1992 and 2009, prescriptions for oral mesalazine increased sixfold, whereas topical suspensions declined by 10%. In clinical trials, topical therapy resulted in higher remission and clinical response rates than oral therapy, with trends to earlier improvement. The mucosal concentrations of 5-ASA achieved by topical agents in the distal colon were up to 200-fold higher than those achieved by oral administration alone. Despite active colitis, over 40% of a topically administered 4 g 5-ASA suspension (equal to 1.6 g) reached the sigmoid colon. This likely represents a therapeutic exposure of 5-ASA. Although topical therapies are less convenient than oral medications, treatment algorithms have failed to take into account quality of life improvements resulting from more rapid and complete treatment response. Topical mesalazine therapy is superior to oral therapy in distal ulcerative colitis for both therapeutic response and drug delivery. Practice patterns should be re-evaluated in light of this information.
Product Description
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Mesalazine, European Pharmacopoeia (EP) Reference Standard