Merck
  • Review article: medication non-adherence in ulcerative colitis--strategies to improve adherence with mesalazine and other maintenance therapies.
Review article: medication non-adherence in ulcerative colitis--strategies to improve adherence with mesalazine and other maintenance therapies.
Alimentary pharmacology & therapeutics (2008-04-04)
A B Hawthorne, G Rubin, S Ghosh
ABSTRACT
Significant number of patients with ulcerative colitis (UC) fail to comply with treatment. To review issues surrounding medication non-adherence in inflammatory bowel disease (IBD), including the clinical and health service implications in the UK, and discuss strategies for optimizing medication adherence. Articles cited were identified via a PubMed search, utilizing the words IBD, adherence, compliance, medication and UC. Medication non-adherence is multifactorial involving factors other than dosing frequency. Male gender (OR: 2.06), new patient status (OR: 2.14), work and travel pressures (OR: 4.9) and shorter disease duration (OR: 2.1), among others are proven predictors of non-adherence in UC. These indicators can identify 'at-risk' patients and allow an individually tailored treatment approach to be introduced that optimizes medication adherence. A collaborative relationship between physician and patient is important; several strategies for improving adherence have been proven effective including open dialogue that takes into consideration the patient's health beliefs and concerns, providing educational (e.g. verbal/written information, self-management programmes) and behavioural interventions (e.g. calendar blister packs, cues/reminders). Educational and behavioural interventions tailored to individual patients can optimize medication adherence. Additional studies combining educational and behavioural interventions may provide further strategies for improving medication adherence rates in UC.
Product Description
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Mesalazine, European Pharmacopoeia (EP) Reference Standard