Merck
  • Estrogen receptor, Progesterone receptor, HER2 status and Ki67 index and responsiveness to adjuvant tamoxifen in postmenopausal high-risk breast cancer patients enrolled in the DBCG 77C trial.
Estrogen receptor, Progesterone receptor, HER2 status and Ki67 index and responsiveness to adjuvant tamoxifen in postmenopausal high-risk breast cancer patients enrolled in the DBCG 77C trial.
European journal of cancer (Oxford, England : 1990) (2014-03-29)
Ann S Knoop, Anne-Vibeke Lænkholm, Maj-Britt Jensen, Kirsten V Nielsen, Jørn Andersen, Dorte Nielsen, Bent Ejlertsen
ABSTRACT
The DBCG 77C trial compared one year of tamoxifen in postmenopausal, steroid-receptor unknown, high-risk breast cancer patients to no adjuvant systemic therapy. After a potential follow-up of 30years we report overall efficacy of the study and results according to subtypes subsequently assessed by immunohistochemistry and fluorescent in situ hybridisation (FISH). Between 1977 and 1982, 1716 postmenopausal patients with tumours larger than 5cm or positive axillary nodes were randomly assigned to no systemic therapy or tamoxifen 30mg daily for one year. Archival tumour tissue from 1515 patients was analysed and the hormone receptor positive (estrogen receptor (ER) and/or progesterone receptor (PR)) cancers were defined as luminal A if Ki67 low and HER2-negative; as luminal B if Ki67 high or HER2-positive; and otherwise as non-luminal-HER2 positive or triple negative. In the intent-to-treat (ITT) population one year of tamoxifen improved the disease-free-survival (DFS) (hazard ratio (HR)=0.87; 95% confidence interval (CI) 0.77-0.98), the Breast Cancer Recurrence Rate (BCRR) (HR=0.79; 0.69-0.90) and reduced the breast-cancer-specific-mortality (BCM) (HR=0.83; 0.73-0.93). BCRR were improved significantly by tamoxifen in luminal A (HR=0.66; 0.53-0.84) and luminal B/HER2- (HR=0.54; 0.39-0.74) but not in the other subsets, and with similar results for BCM with 30years follow-up. One year of treatment with tamoxifen significantly improves BCRR and BCM in postmenopausal patients with ER positive breast cancers. The relative benefit from tamoxifen was not significantly different in luminal A and B subtypes. The Danish Breast Cancer Cooperative Group (DBCG) prepared the original protocol (DBCG 77C) and was the sponsor of the study. Funding was not provided to the participating departments. The biomarker study was supported by grants from the Clinical Institute, Odense University.
Product Description
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Tamoxifen, ≥99%