Merck
  • Nickel oxide nanoparticles induce inflammation and genotoxic effect in lung epithelial cells.
Nickel oxide nanoparticles induce inflammation and genotoxic effect in lung epithelial cells.
Toxicology letters (2014-02-08)
Laura Capasso, Marina Camatini, Maurizio Gualtieri
ABSTRACT
Nickel oxide nanoparticles (NiONPs) toxicity has been evaluated in the human pulmonary epithelial cell lines: BEAS-2B and A549. The nanoparticles, used at the doses of 20, 40, 60, 80, 100 μg/ml, induced a significant reduction of cell viability and an increase of apoptotic and necrotic cells at 24h. A significant release of interleukin-6 and -8 was assessed after 24h of treatment, even intracellular ROS increased already at 45 min after exposure. The results obtained evidenced that the cytokines release was dependent on mitogen activated protein kinases (MAPK) cascade through the induction of NF-kB pathway. NiONPs induced cell cycle alteration in both the cell lines even in different phases and these modifications may be induced by the NPs genotoxic effect, suggested by the nuclear translocation of phospho-ATM and phospho-ATR. Our results confirm the cytotoxic and pro-inflammatory potential of NiONPs. Moreover their ability in inducing DNA damage responses has been demonstrated. Such effects were present in A549 cells which internalize the NPs and BEAS-2B cells in which endocytosis has not been observed.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, foil, 2m coil, thickness 0.015mm, 99.9%
Nickel, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.95%
Nickel, foil, 50x50mm, thickness 0.125mm, as rolled, 99.999%
Nickel, rod, 1000mm, diameter 6.35mm, 99%
Nickel, foil, 25x25mm, thickness 0.3mm, annealed, 99%
Nickel, mesh, 30x30mm, nominal aperture 0.04mm, thickness 0.004mm, wire diameter 0.011mm, 500 wires/inch, open area 60%, electro-formed mesh, 99.9%
Nickel, foil, 50x50mm, thickness 0.3mm, as rolled, 99.999%
Nickel, foil, 25x25mm, thickness 2.0mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, pellets, 50g, max. size 20mm, 99.99+%
Nickel, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%
Nickel, foil, 2m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%
Nickel, mesh, 240x240mm, nominal aperture 0.43mm, thickness 0.004mm, wire diameter 0.067mm, 51 wires/inch, open area 75%, electro-formed mesh, 99.9%
Nickel, foil, 50x50mm, thickness 2.0mm, annealed, 99%
Nickel, foil, 150x300mm, thickness 0.25mm, annealed, 99.98%
Nickel, tube, 500mm, outside diameter 1.5mm, inside diameter 1.4mm, wall thickness 0.05mm, hard, 99.5%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, microfoil, disks, 25mm, thinness 1.0μm, specific density 895.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, foil, 2m coil, thickness 0.02mm, 99.9%
Nickel, wire reel, 10m, diameter 0.125mm, hard, 99.98%
Nickel, foil, 150x300mm, thickness 0.5mm, annealed, 99.98%
Nickel, mesh, 30x30mm, nominal aperture 0.43mm, thickness 0.004mm, wire diameter 0.067mm, 51 wires/inch, open area 75%, electro-formed mesh, 99.9%
Nickel, foil, not light tested, 50x50mm, thickness 0.007mm, 99.95%