Merck
  • Native efflux pumps contribute resistance to antimicrobials of skin and the ability of Staphylococcus aureus to colonize skin.
Native efflux pumps contribute resistance to antimicrobials of skin and the ability of Staphylococcus aureus to colonize skin.
The Journal of infectious diseases (2013-11-28)
Que Chi Truong-Bolduc, Regis A Villet, Zoe A Estabrooks, David C Hooper
ABSTRACT
Staphylococcus aureus colonizes skin in the presence of antimicrobial fatty acids and polyamines. The chromosomally encoded Tet38 efflux transporter confers resistance to tetracycline and fitness in abscesses, but its natural substrates and those of the Nor quinolone efflux pumps are unknown. Susceptibility of tet38 and other pump mutants to and pump gene induction by fatty acids and polyamines were compared. Transport of fatty acids by Tet38 was determined in membrane vesicles. Survival on skin was tested in an adapted mouse skin infection model. The tet38 expression caused a 5- to 8-fold increase in resistance to palmitoleic and undecanoic acids but not polyamines. Subinhibitory concentrations of these fatty acids induced 4-fold increases in tet38 transcripts and competitively inhibited transport of Hoechst 33 342 dye in Tet38 membrane vesicles. Colonization of skin in BALB/c mice was decreased 5-fold in a Δtet38 mutant, which was complemented by plasmid-encoded tet38. Although polyamine minimum inhibitory concentrations (MICs) decreased 4-fold in a norC::cat mutant and increased 8-fold with norC overexpression, spermidine did not induce expression of norC and other pump genes, and norC::cat exhibited wild-type colonization. Antibacterial fatty acids may be natural substrates of Tet38, which contributes to resistance and the ability of S. aureus to colonize skin.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Tetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard