Merck
  • A β-enaminone-initiated multicomponent domino reaction for the synthesis of indoloquinolizines and benzoquinolizines from acyclic precursors.
A β-enaminone-initiated multicomponent domino reaction for the synthesis of indoloquinolizines and benzoquinolizines from acyclic precursors.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-08-16)
Padmakar A Suryavanshi, Vellaisamy Sridharan, J Carlos Menéndez
ABSTRACT
The cerium(IV) ammonium nitrate (CAN)-catalyzed sequential multicomponent reaction between tryptamine, α,β-unsaturated aldehydes, and β-dicarbonyl compounds affords highly substituted indolo[2,3-a]quinolizines in a single synthetic operation. Two rings are generated through the creation of two C-C and two C-N bonds by a domino process comprising initial β-enaminone formation, followed by individual Michael addition, 6-exo-trig cyclization, iminium formation, and Pictet-Spengler steps. Furthermore, the reaction is diastereoselective and affords exclusively compounds with a trans relationship between the H-2 and H-12b protons. The use of amines bearing a less nucleophilic side chain aromatic ring (5-bromotryptamine, 3,4-dimethoxyphenylethylamine) prevents the Pictet-Spengler final step and leads to N-indolylethyl or N-phenylethyl-1,4-dihydropyridines, which are cyclized to the corresponding indolo[2,3-a]quinolizines or benzo[a]quinolizines in the presence of HCl in methanol/water. Treatment of the fused quinolizine derivatives with sodium triacetoxyborohydride led to the corresponding indolo[2,3-a]quinolizidines or benzo[a]quinolizidines, possessing four stereogenic centers, as mixtures of two diastereomers.
Sigma-Aldrich
Ammonium nitrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, SAJ first grade
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%