Merck
Ethylene glycol intoxication: evaluation of kinetics and crystalluria.
The American journal of medicine (1988-01-01)
D Jacobsen, T P Hewlett, R Webb, S T Brown, A T Ordinario, K E McMartin
ABSTRACT
Ethylene glycol and glycolate kinetics were studied in two cases of ethylene glycol intoxication with maximal ethylene glycol/glycolate concentrations of 40.9/26.8 and 56.4/22.4 mmol/liter, respectively. Both patients survived, but with prolonged renal failure, upon treatment with bicarbonate, ethanol, and hemodialysis. Glycolic acid was the major cause of the metabolic acidosis in both cases; lactate levels were only slightly elevated. Kinetic calculations showed that both ethylene glycol and glycolate were distributed in total body water with plasma half-lives of 8.4 and 7.0 hours, respectively. The half-life of ethylene glycol was increased more than 10-fold by ethanol treatment alone. Calcium oxalate monohydrate crystalluria was dominant in both cases, but in one was preceded by a short period with mainly dihydrate excretion; crystalluria was not present upon admission. Repetitive urine microscopy in search of needle- or envelope-shaped crystals should be performed when ethylene glycol intoxication is suspected.
Product Description
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O