Merck
Hematotoxicity in workers exposed to low levels of benzene.
Science (New York, N.Y.) (2004-12-04)
Qing Lan, Luoping Zhang, Guilan Li, Roel Vermeulen, Rona S Weinberg, Mustafa Dosemeci, Stephen M Rappaport, Min Shen, Blanche P Alter, Yongji Wu, William Kopp, Suramya Waidyanatha, Charles Rabkin, Weihong Guo, Stephen Chanock, Richard B Hayes, Martha Linet, Sungkyoon Kim, Songnian Yin, Nathaniel Rothman, Martyn T Smith
ABSTRACT
Benzene is known to have toxic effects on the blood and bone marrow, but its impact at levels below the U.S. occupational standard of 1 part per million (ppm) remains uncertain. In a study of 250 workers exposed to benzene, white blood cell and platelet counts were significantly lower than in 140 controls, even for exposure below 1 ppm in air. Progenitor cell colony formation significantly declined with increasing benzene exposure and was more sensitive to the effects of benzene than was the number of mature blood cells. Two genetic variants in key metabolizing enzymes, myeloperoxidase and NAD(P)H:quinone oxidoreductase, influenced susceptibility to benzene hematotoxicity. Thus, hematotoxicity from exposure to benzene occurred at air levels of 1 ppm or less and may be particularly evident among genetically susceptible subpopulations.
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard