Merck
  • Increased dopamine and its metabolites in SH-SY5Y neuroblastoma cells that express tyrosinase.
Increased dopamine and its metabolites in SH-SY5Y neuroblastoma cells that express tyrosinase.
Journal of neurochemistry (2003-09-27)
Takafumi Hasegawa, Michiko Matsuzaki, Atsushi Takeda, Akio Kikuchi, Katsutoshi Furukawa, Shigeki Shibahara, Yasuto Itoyama
ABSTRACT
Oxidized metabolites of dopamine, known as dopamine quinone derivatives, are thought to play a pivotal role in the degeneration of dopaminergic neurons. Although such quinone derivatives are usually produced via the autoxidation of catecholamines, tyrosinase, which is a key enzyme in melanin biosynthesis via the production of DOPA and subsequent molecules, may potentially accelerate the induction of catecholamine quinone derivatives by its oxidase activity. In the present study, we developed neuronal cell lines in which the expression of human tyrosinase was inducible. Overexpression of tyrosinase in cultured cell lines resulted in (i) increased intracellular dopamine content; (ii) induction of oxidase activity not only for DOPA but also for dopamine; (iii) formation of melanin pigments in cell soma; and (iv) increased intracellular reactive oxygen species. Interestingly, the expressed tyrosinase protein was initially distributed in the entire cytoplasm and then accumulated to form catecholamine-positive granular structures by 3 days after the induction. The granular structures consisted of numerous rounded, dark bodies of melanin pigments and were largely coincident with the distribution of lysosomes. This cellular model that exhibits increased dopamine production will provide a useful tool for detailed analyses of the potentially noxious effects of oxidized catecholamine metabolites.
Product Description
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%