Merck
  • Ionic liquids as superior solvents for headspace gas chromatography of residual solvents with very low vapor pressure, relevant for pharmaceutical final dosage forms.
Ionic liquids as superior solvents for headspace gas chromatography of residual solvents with very low vapor pressure, relevant for pharmaceutical final dosage forms.
Journal of chromatography. A (2009-06-30)
Gerhard Laus, Max Andre, Gino Bentivoglio, Herwig Schottenberger
ABSTRACT
1-n-Butyl-3-methylimidazolium dimethyl phosphate (BMIM DMP) was identified as the most suitable ionic liquid as solvent for the headspace gas chromatographic analysis of solvents with very low vapor pressure such as dimethylsulfoxide, N-methylpyrrolidone, sulfolane, tetralin, and ethylene glycol in a realistic matrix of commonly used excipients (carboxymethylcellulose, magnesium stearate, guar flour, and corn starch) in pharmaceutical products. Limits of quantification and limits of detection were in the low microgram per gram range. The detection of traces of sulfolane in a real sample of tablets containing the drug cefpodoxim proxetil demonstrated the applicability of the method.
Sigma-Aldrich
Sulfolane, 99%
Supelco
Sulfolane, analytical standard
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard
USP
Cefpodoxime proxetil, United States Pharmacopeia (USP) Reference Standard
Supelco
N-Methylpyrrolidone, Pharmaceutical Secondary Standard; Certified Reference Material