Merck
  • Indications for an active process underlying spontaneous and radiation-induced micronucleation in L929 cells.
Indications for an active process underlying spontaneous and radiation-induced micronucleation in L929 cells.
International journal of radiation biology (2000-01-06)
M Abend, S Frombeck, D Van Beuningen
ABSTRACT
To investigate the mechanism of micronucleus formation in irradiated L929 cells. Radiation-induced micronuclei (MN) of L929 cells isolated at 48 and 72 h after irradiation were processed for detection of DNA-laddering and higher-order chromatin fragments using conventional gel electrophoresis and pulse-field gel electrophoresis. Quantification of double-strand breaks in micronuclei and nuclei was performed with the TdT assay and quantified using image analysis. The number of binucleated cells containing micronuclei (cytochalasin B method) was counted after application of three unspecific endonuclease inhibitors (aurin, ATA, spermine), a topoisomerase II inhibitor (VM-26), administration of two PKC inhibitors (H-7, Gö6983) and after addition of N-acetylsphingosine (C2-ceramide). PKC activity was determined by measuring the incorporation of [gamma-32P]ATP into a suitable specific substrate. Proliferation was measured by detection of PCNA, RFP-A and BrdU (30-min pulse labelling) using both conventional immunofluorescence and laser scanning microscopy. (1) Higher chromatin fragments accumulated in MN with a size as they occur during early stages of apoptosis; (2) the frequency of MN was influenced by drugs known to play an important role in signalling and execution of apoptosis (endonucleases, topoisomerase II, protein kinases, ceramide); (3) MN are characterized by a reduced transcription ability (PCNA, RFP-A). A proportion of L929 MN may be formed by an active process comparable with the early stages of apoptosis; it may play a role in the re-organization of the damaged genome.
Sigma-Aldrich
p-Rosolic acid, Dye content 84 %
Sigma-Aldrich
p-Rosolic acid, JIS special grade