Merck
  • Effect of soybean feeding on experimental carcinogenesis--III. Carcinogenecity of nitrite and dibutylamine in mice: a histopathological study.
Effect of soybean feeding on experimental carcinogenesis--III. Carcinogenecity of nitrite and dibutylamine in mice: a histopathological study.
European journal of cancer & clinical oncology (1988-03-01)
N M Mokhtar, A A el-Aaser, M N el-Bolkainy, H A Ibrahim, N K Badr El-Din, N Z Moharram
ABSTRACT
The potential carcinogenic effect of nitrosamine precursors, DBA (dibutylamine) and nitrite, was clearly demonstrated pathologically in the liver and bladder of male Swiss albino mice. Benign tumours were induced in the bladder with an incidence of 40%, and hepatomas were detected in the liver in 27% of the cases. The protective effect of soybean and ascorbic acid, added separately to the diet or to the drinking water respectively, was demonstrated by a marked reduction in dysplastic features and absence of tumour in both the liver and the urinary bladder.
Sigma-Aldrich
Dibutylamine, ≥98%
Sigma-Aldrich
Dibutylamine, puriss., ≥99.0% (GC)
Sigma-Aldrich
Dibutylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Dibutylamine, ≥99.5%