Merck
  • Variation of some inflammatory markers in hypertensive patients after 1 year of olmesartan/amlodipine single-pill combination compared with olmesartan or amlodipine monotherapies.
Variation of some inflammatory markers in hypertensive patients after 1 year of olmesartan/amlodipine single-pill combination compared with olmesartan or amlodipine monotherapies.
Journal of the American Society of Hypertension : JASH (2013-01-17)
Giuseppe Derosa, Arrigo F G Cicero, Anna Carbone, Fabrizio Querci, Elena Fogari, Angela D'Angelo, Pamela Maffioli
ABSTRACT
The purpose of this study was to evaluate a fixed olmesartan/amlodipine combination on blood pressure control, lipid profile, insulin sensitivity, and some inflammatory markers compared with single-drug monotherapy. A total of 276 hypertensive patients were randomly assigned to olmesartan 20 mg, amlodipine 10 mg, or a single pill containing olmesartan/amlodipine combination 20/5 mg for 12 months. We evaluated the following at baseline and after 6 and 12 months: body weight, body mass index, systolic (SBP) and diastolic blood pressures (DBP), fasting plasma glucose (FPG), fasting plasma insulin (FPI), lipid profile, tumor necrosis factor-α (TNF-α), retinol binding protein-4 (RBP-4), and interleukins 6 and 7 (IL-6 and IL-7). At baseline, and after 6 and 12 months, patients underwent an euglycemic, hyperinsulinemic clamp. The olmesartan/amlodipine combination provided a greater decrease of SBP and DPB compared with amlodipine and olmesartan monotherapies. The olmesartan/amlodipine combination decreased FPG after 12 months compared with amlodipine monotherapy. The combination decreased FPI and homeostasis model assessment index and increased M value both compared with baseline and with olmesartan and amlodipine monotherapies. Olmesartan/amlodipine decreased IL-7, but not IL-6, compared with single drug components. The olmesartan/amlodipine combination is effective and safe in reducing blood pressure and has some additive effects not shown by single drugs, such as an improvement of IL-7.
Product Description
Amlodipine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Amlodipine besylate, Pharmaceutical Secondary Standard; Certified Reference Material
Amlodipine besylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)