Merck
  • Reactive oxygen species are generated by the respiratory complex II--evidence for lack of contribution of the reverse electron flow in complex I.
Reactive oxygen species are generated by the respiratory complex II--evidence for lack of contribution of the reverse electron flow in complex I.
The FEBS journal (2012-12-05)
Rafael Moreno-Sánchez, Luz Hernández-Esquivel, Nadia A Rivero-Segura, Alvaro Marín-Hernández, Jiri Neuzil, Stephen J Ralph, Sara Rodríguez-Enríquez
ABSTRACT
Succinate-driven oxidation via complex II (CII) may have a significant contribution towards the high rates of production of reactive oxygen species (ROS) by mitochondria. Here, we show that the CII Q site inhibitor thenoyltrifluoroacetone (TTFA) blocks succinate + rotenone-driven ROS production, whereas the complex III (CIII) Qo inhibitor stigmatellin has no effect, indicating that CII, not CIII, is the ROS-producing site. The complex I (CI) inhibitor rotenone partially reduces the ROS production driven by high succinate levels (5 mm), which is commonly interpreted as being due to inhibition of a reverse electron flow from CII to CI. However, experimental evidence presented here contradicts the model of reverse electron flow. First, ROS levels produced using succinate + rotenone were significantly higher than those produced using glutamate + malate + rotenone. Second, in tumor mitochondria, succinate-driven ROS production was significantly increased (not decreased) by rotenone. Third, in liver mitochondria, rotenone had no effects on succinate-driven ROS production. Fourth, using isolated heart or hepatoma (AS-30D) mitochondria, the CII Qp anti-cancer drug mitochondrially targeted vitamin E succinate (MitoVES) induced elevated ROS production in the presence of low levels of succinate(0.5 mm), but rotenone had no effect. Using sub-mitochondrial particles, the Cu-based anti-cancer drug Casiopeina II-gly enhanced succinate-driven ROS production. Thus, the present results are inconsistent with and question the interpretation of reverse electron flow from CII to CI and the rotenone effect on ROS production supported by succinate oxidation. Instead, a thermodynamically more favorable explanation is that, in the absence of CIII or complex IV (CIV) inhibitors (which, when added, facilitate reverse electron flow by inducing accumulation of ubiquinol, the CI product), the CII redox centers are the major source of succinate-driven ROS production.
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Sigma-Aldrich
2-Thenoyltrifluoroacetone, 99%
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
DL-Malic acid, 99%
Supelco
D-Malic acid, analytical standard