Merck
  • Protease-activated receptor-1 modulates hippocampal memory formation and synaptic plasticity.
Protease-activated receptor-1 modulates hippocampal memory formation and synaptic plasticity.
Journal of neurochemistry (2012-11-02)
Antoine G Almonte, Laura H Qadri, Faraz A Sultan, Jennifer A Watson, Daniel J Mount, Gavin Rumbaugh, J David Sweatt
ABSTRACT
Protease-activated receptor-1 (PAR1) is an unusual G-protein coupled receptor (GPCR) that is activated through proteolytic cleavage by extracellular serine proteases. Although previous work has shown that inhibiting PAR1 activation is neuroprotective in models of ischemia, traumatic injury, and neurotoxicity, surprisingly little is known about PAR1's contribution to normal brain function. Here, we used PAR1-/- mice to investigate the contribution of PAR1 function to memory formation and synaptic function. We demonstrate that PAR1-/- mice have deficits in hippocampus-dependent memory. We also show that while PAR1-/- mice have normal baseline synaptic transmission at Schaffer collateral-CA1 synapses, they exhibit severe deficits in N-methyl-d-aspartate receptor (NMDAR)-dependent long-term potentiation (LTP). Mounting evidence indicates that activation of PAR1 leads to potentiation of NMDAR-mediated responses in CA1 pyramidal cells. Taken together, this evidence and our data suggest an important role for PAR1 function in NMDAR-dependent processes subserving memory formation and synaptic plasticity.
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-NMDAR2A Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NMDAR2B Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-β III tubulin Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 3 Antibody, Chemicon®, from rabbit