Merck
  • Bone quality evaluation: comparison of cone beam computed tomography and subjective surgical assessment.
Bone quality evaluation: comparison of cone beam computed tomography and subjective surgical assessment.
The International journal of oral & maxillofacial implants (2012-10-12)
Jayasankar V Valiyaparambil, Ibrahim Yamany, Denise Ortiz, David M Shafer, David Pendrys, Martin Freilich, Sanjay M Mallya
ABSTRACT
To examine the relationship between dental cone beam computed tomography (CBCT) gray scale values and Hounsfield units (HU), and whether the gray values of edentulous sites correlate with the subjective clinical bone quality assessed at surgery. Two radiographic phantoms containing varying concentrations of either dipotassium hydrogen phosphate or calcium hydroxyapatite (HA) were imaged using multislice CT or CBCT. Reconstructed DICOM data were analyzed to examine the relationship between CBCT gray values and HU. Presurgical CBCT scans from 52 patients who underwent implant placement in the posterior sextants were used. The gray values of the edentulous implant sites were measured and compared with the subjective bone quality assessed at surgery. There was a strong correlation between CBCT gray values and HU. CBCT gray values increased linearly with increasing calcium HA or bone equivalent density material. CBCT gray values measured at edentulous implant sites ranged from -455 to 642, with a trend of decreasing gray values with bone quality type. The median gray values for the four subjective bone types were: 362 (type 1), 214 (type 2), 76 (type 3), and -454 (type 4). CBCT gray values can be used to infer bone density and may provide a valuable aid to predict bone quality at potential implant sites.
Sigma-Aldrich
Phosphorus standard solution, 1000 ppm P
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%