Merck
  • Chromatographic resolution and isotherm determination of (R,S)-mandelic acid on Chiralcel-OD column.
Chromatographic resolution and isotherm determination of (R,S)-mandelic acid on Chiralcel-OD column.
Journal of separation science (2012-07-19)
Shimin Mao, Yan Zhang, Sohrab Rohani, Ajay K Ray
ABSTRACT
Resolution of racemic mandelic acid ((R,S)-MA) and numerical determination of binary competitive isotherm of (R,S)-MA on Chiralcel-OD column have been investigated in this study. The effects of the alcohol modifier and acidic additive in the mobile phase on the retention and enantioseparation of (R,S)-MA were studied at first. The inverse method was then used to determine the competitive isotherm parameters of (R,S)-MA by minimizing the sum of square deviations of the model predictions from the measured elution profiles. The results indicate that the mobile phase with 85% hexane/15% isopropanol/0.3% trifluoroacetic acid mixture gives the best resolution of (R,S)-MA and competitive-modified Langmuir isotherm provides the more accurate sorption mechanism of (R,S)-MA on the cellulose tris(3,5-dimethylphenylcarbamate) chiral stationary phase.
Sigma-Aldrich
(R)-(−)-Mandelic acid, 98%
Sertraline impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(R)-(−)-Mandelic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, ChiPros®, produced by BASF, 99%
Sigma-Aldrich
Mandelic acid, 99%
Supelco
DL-Mandelic acid, analytical standard