Merck
  • Solution-processable pyrite FeS(2) nanocrystals for the fabrication of heterojunction photodiodes with visible to NIR photodetection.
Solution-processable pyrite FeS(2) nanocrystals for the fabrication of heterojunction photodiodes with visible to NIR photodetection.
Advanced materials (Deerfield Beach, Fla.) (2012-06-08)
Di-Yan Wang, You-Ting Jiang, Chih-Cheng Lin, Shao-Sian Li, Yaw-Tyng Wang, Chia-Chun Chen, Chun-Wei Chen
ABSTRACT
A heterojunction photodiode with NIR photoresponse using solution processable pyrite FeS(2) nanocrystal ink is demonstrated which has the advantages of earth-abundance and non-toxicity. The device consists of a FeS(2) nanocrystal (NC) thin film sandwiched with semiconducting metal oxides with a structure of ITO/ZnO/FeS(2) NC/MoO(3) /Au, which exhibits an excellent photoresponse with a spectral response extended to NIR wavelengths of up to 1150 nm and a high photocurrent/dark current ratio of up to 8000 at -1 V under AM1.5 illumination (100 mW cm(-2) ).
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%