Merck
  • Immunotoxicity in ascidians: antifouling compounds alternative to organotins: III--the case of copper(I) and Irgarol 1051.
Immunotoxicity in ascidians: antifouling compounds alternative to organotins: III--the case of copper(I) and Irgarol 1051.
Chemosphere (2012-05-01)
Francesca Cima, Loriano Ballarin
ABSTRACT
After the widespread ban of TBT, due to its severe impact on coastal biocoenoses, mainly related to its immunosuppressive effects on both invertebrates and vertebrates, alternative biocides such as Cu(I) salts and the triazine Irgarol 1051, the latter previously used in agriculture as a herbicide, have been massively introduced in combined formulations for antifouling paints against a wide spectrum of fouling organisms. Using short-term (60 min) haemocyte cultures of the colonial ascidian Botryllus schlosseri exposed to various sublethal concentrations of copper(I) chloride (LC(50)=281 μM, i.e., 17.8 mg Cu L(-1)) and Irgarol 1051 (LC(50)>500 μM, i.e., >127 mg L(-1)), we evaluated their immunotoxic effects through a series of cytochemical assays previously used for organotin compounds. Both compounds can induce dose-dependent immunosuppression, acting on different cellular targets and altering many activities of immunocytes but, unlike TBT, did not have significant effects on cell morphology. Generally, Cu(I) appeared to be more toxic than Irgarol 1051: it significantly (p<0.05) inhibited yeast phagocytosis at 0.1 μM (∼10 μg L(-1)), and affected calcium homeostasis and mitochondrial cytochrome-c oxidase activity at 0.01 μM (∼1 μg L(-1)). Both substances were able to change membrane permeability, induce apoptosis from concentrations of 0.1 μM (∼10 μg L(-1)) and 200 μM (∼50 mg L(-1)) for Cu(I) and Irgarol 1051, respectively, and alter the activity of hydrolases. Both Cu(I) and Irgarol 1051 inhibited the activity of phenoloxidase, but did not show any interactive effect when co-present in the exposure medium, suggesting different mechanisms of action.
Sigma-Aldrich
Copper(II) chloride dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Supelco
Irgarol®, PESTANAL®, analytical standard
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Copper(II) chloride, powder, 99%