Merck
  • Human CD4+ lymphocytes for antigen quantification: characterization using conventional flow cytometry and mass cytometry.
Human CD4+ lymphocytes for antigen quantification: characterization using conventional flow cytometry and mass cytometry.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2012-04-28)
Lili Wang, Fatima Abbasi, Olga Ornatsky, Kenneth D Cole, Martin Misakian, Adolfas K Gaigalas, Hua-Jun He, Gerald E Marti, Scott Tanner, Richard Stebbings
ABSTRACT
To transform the linear fluorescence intensity scale obtained with fluorescent microspheres to an antibody bound per cell (ABC) scale, a biological cell reference material is needed. Optimally, this material should have a reproducible and tight ABC value for the expression of a known clinical reference biomarker. In this study, we characterized commercially available cryopreserved peripheral blood mononuclear cells (PBMCs) and two lyophilized PBMC preparations, Cyto-Trol and PBMC-National Institute for Biological Standard and Control (NIBSC) relative to freshly prepared PBMC and whole blood samples. It was found that the ABC values for CD4 expression on cryopreserved PBMC were consistent with those of freshly obtained PBMC and whole blood samples. By comparison, the ABC value for CD4 expression on Cyto-Trol is lower and the value on PBMC-NIBSC is much lower than those of freshly prepared cell samples using both conventional flow cytometry and CyTOF™ mass cytometry. By performing simultaneous surface and intracellular staining measurements on these two cell samples, we found that both cell membranes are mostly intact. Moreover, CD4(+) cell diameters from both lyophilized cell preparations are smaller than those of PBMC and whole blood. This could result in steric interference in antibody binding to the lyophilized cells. Further investigation of the fixation effect on the detected CD4 expression suggests that the very low ABC value obtained for CD4(+) cells from lyophilized PBMC-NIBSC is largely due to paraformaldehyde fixation; this significantly decreases available antibody binding sites. This study provides confirmation that the results obtained from the newly developed mass cytometry are directly comparable to the results from conventional flow cytometry when both methods are standardized using the same ABC approach.
Sigma-Aldrich
Paraformaldehyde, CP
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Paraformaldehyde, prilled, 95%