Merck
  • Paneth cell ablation in the presence of Klebsiella pneumoniae induces necrotizing enterocolitis (NEC)-like injury in the small intestine of immature mice.
Paneth cell ablation in the presence of Klebsiella pneumoniae induces necrotizing enterocolitis (NEC)-like injury in the small intestine of immature mice.
Disease models & mechanisms (2012-02-14)
Chunxian Zhang, Michael P Sherman, Lawrence S Prince, David Bader, Jörn-Hendrik Weitkamp, James C Slaughter, Steven J McElroy
ABSTRACT
Necrotizing enterocolitis (NEC) is a leading cause of morbidity and mortality in premature infants. During NEC pathogenesis, bacteria are able to penetrate innate immune defenses and invade the intestinal epithelial layer, causing subsequent inflammation and tissue necrosis. Normally, Paneth cells appear in the intestinal crypts during the first trimester of human pregnancy. Paneth cells constitute a major component of the innate immune system by producing multiple antimicrobial peptides and proinflammatory mediators. To better understand the possible role of Paneth cell disruption in NEC, we quantified the number of Paneth cells present in infants with NEC and found that they were significantly decreased compared with age-matched controls. We were able to model this loss in the intestine of postnatal day (P)14-P16 (immature) mice by treating them with the zinc chelator dithizone. Intestines from dithizone-treated animals retained approximately half the number of Paneth cells compared with controls. Furthermore, by combining dithizone treatment with exposure to Klebsiella pneumoniae, we were able to induce intestinal injury and inflammatory induction that resembles human NEC. Additionally, this novel Paneth cell ablation model produces NEC-like pathology that is consistent with other currently used animal models, but this technique is simpler to use, can be used in older animals that have been dam fed, and represents a novel line of investigation to study NEC pathogenesis and treatment.
Sigma-Aldrich
Dithizone, ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)
Sigma-Aldrich
Dithizone, Practical Grade
Sigma-Aldrich
Dithizone, JIS special grade, ≥85.0%