Merck
Arbutin derivatives from the seeds of Madhuca latifolia.
Natural product communications (2012-01-10)
Shazia Khan, M Nadeem Kardar, Bina S Siddiqui
ABSTRACT
A new arbutin derivative, madhuglucoside (1), along with three known arbutin derivatives were isolated from the seeds of Madhuca latifolia in addition to seven other known constituents. Their structures were established on the basis of spectral analysis. Compounds 1a, 2a and 3a were obtained in a pure state after acetylation of the mother fraction and characterized as their acetyl derivatives.
Sigma-Aldrich
Arbutin, ≥98% (HPLC)
Arbutin, European Pharmacopoeia (EP) Reference Standard
Supelco
Arbutin, analytical standard