Merck
  • Surface modification of gadolinium oxide thin films and nanoparticles using poly(ethylene glycol)-phosphate.
Surface modification of gadolinium oxide thin films and nanoparticles using poly(ethylene glycol)-phosphate.
Langmuir : the ACS journal of surfaces and colloids (2011-10-06)
Andrée-Anne Guay-Bégin, Pascale Chevallier, Luc Faucher, Stéphane Turgeon, Marc-André Fortin
ABSTRACT
The performance of nanomaterials for biomedical applications is highly dependent on the nature and the quality of surface coatings. In particular, the development of functionalized nanoparticles for magnetic resonance imaging (MRI) requires the grafting of hydrophilic, nonimmunogenic, and biocompatible polymers such as poly(ethylene glycol) (PEG). Attached at the surface of nanoparticles, this polymer enhances the steric repulsion and therefore the stability of the colloids. In this study, phosphate molecules were used as an alternative to silanes or carboxylic acids, to graft PEG at the surface of ultrasmall gadolinium oxide nanoparticles (US-Gd(2)O(3), 2-3 nm diameter). This emerging, high-sensitivity "positive" contrast agent is used for signal enhancement in T(1)-weighted molecular and cellular MRI. Comparative grafting assays were performed on Gd(2)O(3) thin films, which demonstrated the strong reaction of phosphate with Gd(2)O(3) compared to silane and carboxyl groups. Therefore, PEG-phosphate was preferentially used to coat US-Gd(2)O(3) nanoparticles. The grafting of this polymer on the particles was confirmed by XPS and FTIR. These analyses also demonstrated the strong attachment of PEG-phosphate at the surface of Gd(2)O(3), forming a protective layer on the nanoparticles. The stability in aqueous solution, the relaxometric properties, and the MRI signal of PEG-phosphate-covered Gd(2)O(3) particles were also better than those from non-PEGylated nanoparticles. As a result, reacting PEG-phosphate with Gd(2)O(3) particles is a promising, rapid, one-step procedure to PEGylate US-Gd(2)O(3) nanoparticles, an emerging "positive" contrast agent for preclinical molecular and cellular applications.
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%