Merck
  • [Effectiveness of social mobilization and social marketing in promoting NaFeEDTA-fortified soya sauce in adult women].
[Effectiveness of social mobilization and social marketing in promoting NaFeEDTA-fortified soya sauce in adult women].
Wei sheng yan jiu = Journal of hygiene research (2011-06-24)
Bo Wang, Junshi Chen, Siyan Zhan, Jing Sun, Liming Li
ABSTRACT
To assess the effectiveness of social mobilization and social marketing in promoting NaFeEDTA-fortified soy sauce in an iron deficient population. This study was an uncontrolled, community-based, before-after study, which was implemented in three counties of Shijiazhuang Municipality. The intervention was a social mobilization and social marketing strategy. Adult women older than 20 years of age participated in the evaluation protocol. The main outcomes included KAP relevant to IDA. Cross-sectional samples were used to assess the outcomes at baseline and 1 year later. Knowledge and attitudes of adult women had changed positively, and the percentage of women who had adopted NaFeEDTA-fortified soy sauce increased from 8.9% to 36.6% (P < 0.001). Social mobilization and social marketing had a positive impact on the KAP of adult women in the iron deficient population.
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, SAJ special grade, ≥95.0%