Merck
  • Effects of chloroacetaldehyde in 2-chloroethanol-induced cardiotoxicity.
Effects of chloroacetaldehyde in 2-chloroethanol-induced cardiotoxicity.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-01-27)
Yng-Tay Chen, Ching-I Hsu, Dong-Zong Hung, Isao Matsuura, Jiunn-Wang Liao
ABSTRACT
Cardiovascular effects have often been found in 2-chloroethanol (2-CE) intoxicated patients, but the 2-CE elicits cardiovascular toxicity mechanism is not clear. Recently, we have found that chloroacetaldehyde (CAA) accumulation in 2-CE-intoxicated rat's blood and play an important role in 2-CE intoxication. In this study, we used an isolated rat atrium model to examine the cardiotoxicity of 2-CE and CAA. Results indicated that 2-CE did not cause tension arrest in isolated rat right atria, but CAA did. 2-CE caused tension inhibition in the isolated rat left atria. In addition, CAA caused significant tension inhibition and contracture in the isolated rat left atria. Nifedipine, an L-type calcium channel blocker, decreased CAA-induced tension inhibition and contracture. Meanwhile, atrial nNOS and calmodulin (CaM) had significantly greater expression in the 2-CE group and the CAA group than control group. Nifedipine could decrease CAA-induced nNOS and CaM expression. 2-CE-induced cardiovascular toxicity might be due to its metabolite CAA. CAA-induced cardiovascular toxicity might be mediated by calcium channel and nifedipine protected against nNOS-triggered cardiovascular effects.
Sigma-Aldrich
Chloroacetaldehyde solution, ~55 wt. % in H2O
Sigma-Aldrich
Chloroacetaldehyde solution, Wacker Chemie AG, ≥45.0% in H2O (density determination)
Sigma-Aldrich
2-Chloroethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Chloroethanol, 99%