Merck
  • Application of gamma irradiation in ginseng for both photodegradation of pesticide pentachloronitrobenzene and microbial decontamination.
Application of gamma irradiation in ginseng for both photodegradation of pesticide pentachloronitrobenzene and microbial decontamination.
Journal of hazardous materials (2009-12-05)
Hsiao-Wei Wen, Ming-Fa Hsieh, Ya-Ting Wang, Hsiao-Ping Chung, Po-Chow Hsieh, I-Hsin Lin, Fong-In Chou
ABSTRACT
This study investigates the feasibility of using gamma irradiation for photodegradation of a common residual fungicide, pentachloronitrobenzene (PCNB), in ginseng, and for microbial decontamination. American ginseng, Panax quinquefolius, was subjected to gamma irradiation. PCNB residues were analyzed by gas chromatography with electron capture detection and mass spectrometry. Eighty percent of PCNB (100 ppm) in a methanol aqueous solution was degraded by 5 kGy irradiation, and the primary degradation product was pentachloroaniline. Furthermore, contaminated PCNB (3.7 ppm) in ginseng were reduced to 0.2 ppm after 20 kGy irradiation. The IC(50) for treatment of Sclerotium rolfsii with 20 kGy irradiated PCNB was about 2.7 times higher than that for treatment with unirradiated PCNB. The survival rate of mouse fibroblast L929 cells treated with 20 kGy irradiated PCNB was about 12.9% higher than that of L929 cells treated with unirradiated PCNB. Additionally, after 20 kGy irradiation, less than 5% reduction of contents of ginsenoside Rb1 and Re were observed, and amounts of ginsenosides Rc, Rd, and Rg1 were not reduced significantly. The minimal gamma dose for microbial decontamination was 10 kGy. Therefore, gamma irradiation can be used for both PCNB photodegradation and microbial decontamination of ginseng without obvious loses of ginsenoside contents.
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%
Supelco
Pentachloroaniline, PESTANAL®, analytical standard
Supelco
Quintozene, PESTANAL®, analytical standard
Supelco
Pentachloronitrobenzene solution, certified reference material, 5000 μg/mL in methanol