Merck
  • Treatment with potassium bicarbonate lowers calcium excretion and bone resorption in older men and women.
Treatment with potassium bicarbonate lowers calcium excretion and bone resorption in older men and women.
The Journal of clinical endocrinology and metabolism (2008-10-23)
Bess Dawson-Hughes, Susan S Harris, Nancy J Palermo, Carmen Castaneda-Sceppa, Helen M Rasmussen, Gerard E Dallal
ABSTRACT
Bicarbonate has been implicated in bone health in older subjects on acid-producing diets in short-term studies. The objective of this study was to determine the effects of potassium bicarbonate and its components on changes in bone resorption and calcium excretion over 3 months in older men and women. In this double-blind, controlled trial, 171 men and women age 50 and older were randomized to receive placebo or 67.5 mmol/d of potassium bicarbonate, sodium bicarbonate, or potassium chloride for 3 months. All subjects received calcium (600 mg of calcium as triphosphate) and 525 IU of vitamin D(3) daily. Twenty-four-hour urinary N-telopeptide and calcium were measured at entry and after 3 months. Changes in these measures were compared across treatment groups in the 162 participants included in the analyses. Bicarbonate affected the study outcomes, whereas potassium did not; the two bicarbonate groups and the two no bicarbonate groups were therefore combined. Subjects taking bicarbonate had significant reductions in urinary N-telopeptide and calcium excretion, when compared with subjects taking no bicarbonate (both before and after adjustment for baseline laboratory value, sex, and changes in urinary sodium and potassium; P = 0.001 for both, adjusted). Potassium supplementation did not significantly affect N-telopeptide or calcium excretion. Bicarbonate, but not potassium, had a favorable effect on bone resorption and calcium excretion. This suggests that increasing the alkali content of the diet may attenuate bone loss in healthy older adults.
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, for inorganic trace analysis, 99.7-100.5% dry basis
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)