Merck
  • An environmental approach to correcting iodine deficiency: supplementing iodine in soil by iodination of irrigation water in remote areas.
An environmental approach to correcting iodine deficiency: supplementing iodine in soil by iodination of irrigation water in remote areas.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2008-03-06)
Qiang Ren, Jie Fan, Zhizhong Zhang, Xiaoying Zheng, G Robert Delong
ABSTRACT
Iodine deficiency disorders continue to be a severe problem in many parts of Central Asia, causing delayed mental development and cretinism in indigenous populations. In some areas, iodized salt has not succeeded in controlling this problem. In southern Xinjiang Province of China, we tried a new method of supplying iodine to rural populations by dripping potassium iodate into irrigation water canals. By this means iodine was distributed into soil, crops, animals and people. This proved feasible and cost effective; it reached all the people, required no medical expertise, required no continuing effort after the initial dripping, and had the important added benefit of improving livestock production. We serially monitored iodine concentrations in soil, crops, animal products and human urine for several years after the last dripping. In a similar project in Inner Mongolia, total soil iodine was determined in addition. Here, iodine concentrations in soil, crops, animals and people have been monitored for 4 years after supplementation. After dripping, total iodine increased two-fold, while soluble iodine increased 4-5-fold. Iodine added to soil is available for more than 4 years after a single application. Potassium iodate added to soil appears to increase soluble iodine out of proportion to the amount added. This effect and the long persistence of dripped iodate in soil contribute to the efficacy and cost effectiveness of this method of iodine supplementation.
Product Description
Supelco
Potassium iodate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium iodate solution, 1/60 M
Sigma-Aldrich
Potassium iodate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium iodate, 99.995% trace metals basis
Sigma-Aldrich
Potassium iodate, ACS reagent, 99.5%
Sigma-Aldrich
Potassium iodate, reagent grade, ≥98%