Merck
An unusual cause of neonatal coagulopathy and liver disease.
Journal of perinatology : official journal of the California Perinatal Association (2007-04-25)
C Dryden, J H Simpson, L E Hunter, L Jackson
ABSTRACT
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital viral infection with co-existent hepatic dysfunction and coagulopathy. The associated hepatic dysfunction was so severe that liver biopsy was scheduled before the diagnosis emerged. A high degree of clinical suspicion coupled with prenatal identification of pregnancies at risk of complication by congenital thyrotoxicosis is imperative to facilitate prompt diagnosis and treatment.
Product Description
Supelco
Potassium iodate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium iodate solution, 1/60 M
Sigma-Aldrich
Potassium iodate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium iodate, 99.995% trace metals basis
Sigma-Aldrich
Potassium iodate, ACS reagent, 99.5%
Sigma-Aldrich
Potassium iodate, reagent grade, ≥98%