Merck
Concise asymmetric total synthesis of obolactone.
The Journal of organic chemistry (2006-03-25)
Jiyong Zhang, Yang Li, Wenkuan Wang, Xuegong She, Xinfu Pan
ABSTRACT
The first efficient asymmetric synthesis of obolactone 1 has been accomplished in 11 steps and with a 15% overall yield in which Brown's enantioselective allylation reactions and ring-closing metathesis reaction are key steps.
Sigma-Aldrich
4-Phenyl-3-buten-2-one, 99%
Sigma-Aldrich
trans-4-Phenyl-3-buten-2-one, ≥99%
Sigma-Aldrich
Benzylideneacetone, ≥98%, FG