Merck
  • Contribution of phospholipid pyrrolization to the color reversion produced during deodorization of poorly degummed vegetable oils.
Contribution of phospholipid pyrrolization to the color reversion produced during deodorization of poorly degummed vegetable oils.
Journal of agricultural and food chemistry (2004-06-24)
Rosario Zamora, Carmen Olmo, José L Navarro, Francisco J Hidalgo
ABSTRACT
The Ehrlich reaction was optimized to determine the formation of pyrrolized phospholipids in edible oils in an attempt to understand the color reversion produced during the deodorization of poorly degummed edible oils. The procedure consisted of the treatment of the oil with p-(dimethylamino)benzaldehyde in tetrahydrofuran/2-propanol at a controlled acidity and temperature and the spectrophotometric determination of adducts produced. The extinction coefficient of Ehrlich adducts was calculated by using 1-[1-(2-hydroxyethyl)-1H-pyrrol-2-yl]propan-1-ol (1) as a standard and was 15 300 M(-)(1) cm(-)(1). The response was linear and reproducible within the range of 0.334-48.6 microM of compound 1. When the assay was applied to a soybean oil treated with 100-1000 ppm of phosphatidylethanolamine and submitted to deodorization, the formation of pyrrolized phospholipids was observed at the same time that the disappearance of the phospholipid and the oil darkening were produced. The main changes were observed during the first steps of the deodorization process, when the oil was heated between 80 and 160 degrees C. During the initial heating of the oil until achieving 200 degrees C, oil darkening, phosphatidylethanolamine disappearance, and pyrrolized phospholipid formation were correlated, therefore suggesting a contribution of phospholipid pyrrolization to the oil darkening produced.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, JIS special grade, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Supelco
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Millipore
Kovac′s reagent for indoles, suitable for microbiology