Merck
Polyoma virus nephropathy, first reported case in Saudi Arabia.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2006-08-15)
N A Siddiqi, M H Hamid, A El-Tayeb, E Bokhari
ABSTRACT
Polyoma virus nephropathy (BK virus) is being recognized as an important cause of graft failure. It is usually confused with acute rejection. No cases have been reported from the kingdom of Saudi Arabia. We report a case of a Saudi gentleman, who was transplanted outside the country, with persistently elevated creatinine and urethral stenosis. He was treated for acute rejection on more than one occasion with no significant improvement in his renal function. Polyoma virus nephropathy was diagnosed by detecting the virus DNA by the Poly chain reaction technique (PCR). The patient's renal function stabilized after the calcineurin inhibitors were discontinued.
Sigma-Aldrich
Creatinase from Pseudomonas sp., recombinant, expressed in E. coli, lyophilized powder, 10-15 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Creatinase from microorganisms, lyophilized powder, ≥4 units/mg solid