Merck
  • An amperometric enzyme electrode for creatine determination prepared by the immobilization of creatinase and sarcosine oxidase in poly(vinylferrocenium).
An amperometric enzyme electrode for creatine determination prepared by the immobilization of creatinase and sarcosine oxidase in poly(vinylferrocenium).
Artificial cells, blood substitutes, and immobilization biotechnology (2006-03-16)
Pinar Esra Erden, Sule Pekyardimci, Esma Kiliç, Fatma Arslan
ABSTRACT
A new enzyme electrode for the determination of creatine was developed by immobilizing creatinase (CI) and sarcosine oxidase (SO). The enzymes were co-immobilized in a poly(vinylferrocenium) matrix onto the surface of a platinum working electrode. Crosslinking with glutaraldehyte (GA) and bovine serum albumin (BSA) was selected as the best immobilization method for the enzymatic system. Determination of creatine was performed by the oxidation of enzymatically generated H2O2 at + 0.7 V vs. Ag/AgCl. The linear working range of the electrode was 2.0 x 10(-5) - 3.2 x 10(-4) M and the response time was about 50 s. The effects of pH, temperature, enzyme ratio and buffer concentration were investigated and optimum parameters were found to be 7.5, 37 degrees C, 2.5:1 (CI:SO) and 0.05 M, respectively. The stability and reproducibility of the enzyme electrode have been also studied.
Sigma-Aldrich
Creatinase from Pseudomonas sp., recombinant, expressed in E. coli, lyophilized powder, 10-15 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Creatinase from microorganisms, lyophilized powder, ≥4 units/mg solid
Pricing and availability is not currently available.