Merck
  • JMJD6 Shapes a Pro-tumor Microenvironment via ANXA1-Dependent Macrophage Polarization in Breast Cancer.
JMJD6 Shapes a Pro-tumor Microenvironment via ANXA1-Dependent Macrophage Polarization in Breast Cancer.
Molecular cancer research : MCR (2023-03-04)
Bianca Cioni, Silvia Ratti, Annamaria Piva, Irene Tripodi, Matteo Milani, Francesca Menichetti, Tiziana Langella, Laura Botti, Loris De Cecco, Claudia Chiodoni, Daniele Lecis, Mario P Colombo
ABSTRACT
Breast cancer is the most common type of cancer in women worldwide, with the luminal subtype being the most widespread. Although characterized by better prognosis compared with other subtypes, luminal breast cancer is still considered a threatening disease due to therapy resistance, which occurs via both cell- and non-cell-autonomous mechanisms. Jumonji domain-containing 6, arginine demethylase and lysine hydroxylase (JMJD6) is endowed with a negative prognostic value in luminal breast cancer and, via its epigenetic activity, it is known to regulate many intrinsic cancer cell pathways. So far, the effect of JMJD6 in molding the surrounding microenvironment has not been explored.Here, we describe a novel function of JMJD6 showing that its genetic inhibition in breast cancer cells suppresses lipid droplet formation and ANXA1 expression, via estrogen receptor alpha and PPARα modulation. Reduction of intracellular ANXA1 results in decreased release in the tumor microenvironment (TME), ultimately preventing M2-type macrophage polarization and tumor aggressiveness. Our findings identify JMJD6 as a determinant of breast cancer aggressiveness and provide the rationale for the development of inhibitory molecules to reduce disease progression also through the remodeling of TME composition.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phosphatidylserine Receptor antibody produced in rabbit, 1-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid