Merck
  • Neuronal ER-plasma membrane junctions couple excitation to Ca2+-activated PKA signaling.
Neuronal ER-plasma membrane junctions couple excitation to Ca2+-activated PKA signaling.
Nature communications (2023-08-27)
Nicholas C Vierra, Luisa Ribeiro-Silva, Michael Kirmiz, Deborah van der List, Pradeep Bhandari, Olivia A Mack, James Carroll, Elodie Le Monnier, Sue A Aicher, Ryuichi Shigemoto, James S Trimmer
ABSTRACT
Junctions between the endoplasmic reticulum (ER) and the plasma membrane (PM) are specialized membrane contacts ubiquitous in eukaryotic cells. Concentration of intracellular signaling machinery near ER-PM junctions allows these domains to serve critical roles in lipid and Ca2+ signaling and homeostasis. Subcellular compartmentalization of protein kinase A (PKA) signaling also regulates essential cellular functions, however, no specific association between PKA and ER-PM junctional domains is known. Here, we show that in brain neurons type I PKA is directed to Kv2.1 channel-dependent ER-PM junctional domains via SPHKAP, a type I PKA-specific anchoring protein. SPHKAP association with type I PKA regulatory subunit RI and ER-resident VAP proteins results in the concentration of type I PKA between stacked ER cisternae associated with ER-PM junctions. This ER-associated PKA signalosome enables reciprocal regulation between PKA and Ca2+ signaling machinery to support Ca2+ influx and excitation-transcription coupling. These data reveal that neuronal ER-PM junctions support a receptor-independent form of PKA signaling driven by membrane depolarization and intracellular Ca2+, allowing conversion of information encoded in electrical signals into biochemical changes universally recognized throughout the cell.
Sigma-Aldrich
1-β-D-Arabinofuranosylcytosine, Anticancer, antiviral agent that is especially effective against leukemias.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%