Merck
  • Perfluorooctane Sulfonic Acid Disrupts Protective Tight Junction Proteins via Protein Kinase D in Airway Epithelial Cells.
Perfluorooctane Sulfonic Acid Disrupts Protective Tight Junction Proteins via Protein Kinase D in Airway Epithelial Cells.
Toxicological sciences : an official journal of the Society of Toxicology (2022-09-16)
Joseph H Lucas, Qixin Wang, Irfan Rahman
ABSTRACT
Perfluorooctane sulfonic acid (PFOS) is a long chain per- and polyfluoroalklyl substance (PFAS) that has been used in aqueous film-forming foams. Emerging epidemiological evidence indicates that PFOS may be associated with chronic lung diseases such as asthma and analysis of human tissues demonstrates that the lungs carry a significant body burden of PFOS. Deficits in barrier function are a major risk factor for asthma. Thus, we hypothesized that PFOS exposure will lead to impaired epithelial barrier function through dysregulated tight junctions. Hence, we assessed the impact of PFOS on epithelial barrier integrity. Bronchial epithelial cells (16HBE) were grown on collagen-coated transwells and treated to 5-25 μM PFOS, and assessed for changes in barrier function and tight junction proteins. Rescue experiments were performed using the protein kinase D (PKD) inhibitor, CID755673. PFOS treatment reduced transepithelial electrical resistance (TEER) and increased 4 kDa FITC-dextran flux. Additionally, PFOS significantly decreased protein levels and the tight junction organization rate of occludin and zonula occludens 1. Increased phosphorylation (Ser744/Ser748) of PKD was observed 3 h following PFOS treatment. Pretreatment with the PKD inhibitor attenuated PFOS-mediated changes in TEER and FITC-dextran flux and restored occludin protein levels. In conclusion, PFOS causes loss of airway barrier integrity and the disruption of tight junctions in bronchial epithelial cells, which was partly attenuated through the inhibition of PKD. These findings demonstrate that PFOS is capable of disrupting airway barrier function, a potentially driving factor underlying associations between PFOS and respiratory diseases such as asthma.
Sigma-Aldrich
16HBE14o- Human Bronchial Epithelial Cell Line, 16HBE14o- human bronchial epithelial cell line is widely used to model barrier function of the airway epithelium and to study respiratory ion transport as well as the function of CFTR.
Sigma-Aldrich
PKD Inhibitor, CID755673, The PKD Inhibitor, CID755673, also referenced under CAS 521937-07-5, controls the biological activity of PKD. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.