Merck
  • A kinase-independent function of cyclin-dependent kinase 6 promotes outer radial glia expansion and neocortical folding.
A kinase-independent function of cyclin-dependent kinase 6 promotes outer radial glia expansion and neocortical folding.
Proceedings of the National Academy of Sciences of the United States of America (2022-09-13)
Lei Wang, Jun Young Park, Fengming Liu, Kris M Olesen, Shirui Hou, Jamy C Peng, Jordan Infield, Anna C Levesque, Yong-Dong Wang, Hongjian Jin, Yiping Fan, Patrick J Connelly, Shondra M Pruett-Miller, Miaofen G Hu, Philip W Hinds, Young-Goo Han
ABSTRACT
The neocortex, the center for higher brain function, first emerged in mammals and has become massively expanded and folded in humans, constituting almost half the volume of the human brain. Primary microcephaly, a developmental disorder in which the brain is smaller than normal at birth, results mainly from there being fewer neurons in the neocortex because of defects in neural progenitor cells (NPCs). Outer radial glia (oRGs), NPCs that are abundant in gyrencephalic species but rare in lissencephalic species, are thought to play key roles in the expansion and folding of the neocortex. However, how oRGs expand, whether they are necessary for neocortical folding, and whether defects in oRGs cause microcephaly remain important questions in the study of brain development, evolution, and disease. Here, we show that oRG expansion in mice, ferrets, and human cerebral organoids requires cyclin-dependent kinase 6 (CDK6), the mutation of which causes primary microcephaly via an unknown mechanism. In a mouse model in which increased Hedgehog signaling expands oRGs and intermediate progenitor cells and induces neocortical folding, CDK6 loss selectively decreased oRGs and abolished neocortical folding. Remarkably, this function of CDK6 in oRG expansion did not require its kinase activity, was not shared by the highly similar CDK4 and CDK2, and was disrupted by the mutation causing microcephaly. Therefore, our results indicate that CDK6 is conserved to promote oRG expansion, that oRGs are necessary for neocortical folding, and that defects in oRG expansion may cause primary microcephaly.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Radial Glial Cell Marker-2 Antibody, clone RC2, clone RC2, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Anti-SATB2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Tbr1 Antibody, from chicken, purified by affinity chromatography