Merck
Why Great Mitotic Inhibitors Make Poor Cancer Drugs.
Trends in cancer (2020-06-17)
Victoria C Yan, Hannah E Butterfield, Anton H Poral, Matthew J Yan, Kristine L Yang, Cong-Dat Pham, Florian L Muller
ABSTRACT
Chemotherapy is central to oncology, perceived to operate only on prolific cancerous tissue. Yet, many non-neoplastic tissues are more prolific compared with typical tumors. Chemotherapies achieve sufficient therapeutic windows to exert antineoplastic activity because they are prodrugs that are bioactivated in cancer-specific environments. The advent of precision medicine has obscured this concept, favoring the development of high-potency kinase inhibitors. Inhibitors of essential mitotic kinases exemplify this paradigm shift, but intolerable on-target toxicities in more prolific normal tissues have led to repeated failures in the clinic. Proliferation rates alone cannot be used to achieve cancer specificity. Here, we discuss integrating the cancer specificity of prodrugs from classical chemotherapeutics and the potency of mitotic kinase inhibitors to generate a class of high-precision cancer therapeutics.
Sigma-Aldrich
Anti-TYR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PON2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution