Merck
  • Functional characterization of the ectopically expressed olfactory receptor 2AT4 in human myelogenous leukemia.
Functional characterization of the ectopically expressed olfactory receptor 2AT4 in human myelogenous leukemia.
Cell death discovery (2016-08-24)
S Manteniotis, S Wojcik, P Brauhoff, M Möllmann, L Petersen, J R Göthert, W Schmiegel, U Dührsen, G Gisselmann, H Hatt
ABSTRACT
The olfactory receptor (OR) family was found to be expressed mainly in the nasal epithelium. In the last two decades members of the OR family were detected to be functional expressed in different parts of the human body such as in liver, prostate or intestine cancer cells. Here, we detected the expression of several ORs in the human chronic myelogenous leukemia (CML) cell line K562 and in white blood cells of clinically diagnosed acute myeloid leukemia (AML) patients by RT-PCR and next-generation sequencing. With calcium-imaging, we characterized in greater detail the cell biological role of one OR (OR2AT4) in leukemia. In both cell systems, the OR2AT4 agonist Sandalore-evoked strong Ca(2+) influx via the adenylate cyclase-cAMP-mediated pathway. The OR2AT4 antagonist Phenirat prevented the Sandalore-induced intracellular Ca(2+) increase. Western blot and flow cytometric experiments revealed that stimulation of OR2AT4 reduced the proliferation by decreasing p38-MAPK phosphorylation and induced apoptosis via phosphorylation of p44/42-MAPK. Furthermore, Sandalore increased the number of hemoglobin-containing cells in culture. We described for the first time an OR-mediated pathway in CML and AML that can regulate proliferation, apoptosis and differentiation after activation. This mechanism offers novel therapeutic options for the treatment of AML.
Sigma-Aldrich
HDL mimetic peptide 4F trifluoroacetate, ≥95% (HPLC)
Sigma-Aldrich
3-Methyl-5-(2,2,3-trimethyl-3-cyclopentenyl)pentan-2-ol, ≥90% (GC)