Merck
  • The Allergen Der p3 from House Dust Mite Stimulates Store-Operated Ca2+ Channels and Mast Cell Migration through PAR4 Receptors.
The Allergen Der p3 from House Dust Mite Stimulates Store-Operated Ca2+ Channels and Mast Cell Migration through PAR4 Receptors.
Molecular cell (2018-04-21)
Yu-Ping Lin, Charmaine Nelson, Holger Kramer, Anant B Parekh
ABSTRACT
The house dust mite is the principal source of perennial aeroallergens in man. How these allergens activate innate and adaptive immunity is unclear, and therefore, there are no therapies targeting mite allergens. Here, we show that house dust mite extract activates store-operated Ca2+ channels, a common signaling module in numerous cell types in the lung. Activation of channel pore-forming Orai1 subunits by mite extract requires gating by STIM1 proteins. Although mite extract stimulates both protease-activated receptor type 2 (PAR2) and PAR4 receptors, Ca2+ influx is more tightly coupled to the PAR4 pathway. We identify a major role for the serine protease allergen Der p3 in stimulating Orai1 channels and show that a therapy involving sub-maximal inhibition of both Der p3 and Orai1 channels suppresses mast cell activation to house dust mite. Our results reveal Der p3 as an important aeroallergen that activates Ca2+ channels and suggest a therapeutic strategy for treating mite-induced asthma.
Sigma-Aldrich
Nafamostat mesylate, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Camostat mesylate, ≥98% (HPLC)
Sigma-Aldrich
E-64, protease inhibitor