Merck
  • Recent applications of click chemistry for the functionalization of gold nanoparticles and their conversion to glyco-gold nanoparticles.
Recent applications of click chemistry for the functionalization of gold nanoparticles and their conversion to glyco-gold nanoparticles.
Beilstein journal of organic chemistry (2018-01-31)
Vivek Poonthiyil, Thisbe K Lindhorst, Vladimir B Golovko, Antony J Fairbanks
ABSTRACT
Glycoscience, despite its myriad of challenges, promises to unravel the causes of, potential new detection methods for, and novel therapeutic strategies against, many disease states. In the last two decades, glyco-gold nanoparticles have emerged as one of several potential new tools for glycoscientists. Glyco-gold nanoparticles consist of the unique structural combination of a gold nanoparticle core and an outer-shell comprising multivalent presentation of carbohydrates. The combination of the distinctive physicochemical properties of the gold core and the biological function/activity of the carbohydrates makes glyco-gold nanoparticles a valuable tool in glycoscience. In this review we present recent advances made in the use of one type of click chemistry, namely the azide-alkyne Huisgen cycloaddition, for the functionalization of gold nanoparticles and their conversion to glyco-gold nanoparticles.
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 90 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 50 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 60 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 80 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 70 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 100 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, azide functionalized, 1 ML, OD 50, in water