- Multiresidue procedure to assess the occurrence and dissipation of fungicides and insecticides in vineyard soils from Northwest Spain.
Multiresidue procedure to assess the occurrence and dissipation of fungicides and insecticides in vineyard soils from Northwest Spain.
The presence of fungicide and insecticide residues in wine has been largely investigated. However, few studies have addressed the persistence of these compounds in vineyard soils. In this research, we investigate the residues of a relevant number of fungicides and insecticides in vineyard soils, obtained in the Northwest of Spain, at the beginning of each agriculture campaign. Moreover, the dissipation of species showing high concentrations were monitored during the non-vegetative period of vines, in order to understand their soil evolution between application campaigns. To this end, a multiresidue analytical procedure based on pressurized liquid extraction (PLE) followed by liquid chromatography tandem mass spectrometry (LC-MS/MS) determination was first optimized. Under final working conditions, absolute recoveries in the range from 70 to 130% were achieved for 44 out of 51 selected compounds. The method LOQs remained at the low ng g-1 level (0.2-13 ng g-1) with a linear response range up to 500 ng g-1. Analysis of vineyard soils, collected during a 2-year period, from a geographic area with a high incidence of fungal diseases, demonstrated the presence of relevant concentrations of several fungicides and the insecticide imidacloprid (IMI) in this compartment. Most compounds detected at the end of the application season remained in soil at the beginning of the next year campaign. Among them, six fungicides (dimethomorph, boscalid, myclobutanil, penconazole, pyraclostrobin and pyrimethanil) and IMI showed average dissipation efficiencies below 50%, so they pose a potential to accumulate in this kind of soils.