Merck
A tetra(L-lysine)-grafted poly(organophosphazene) for gene delivery.
Bioorganic & medicinal chemistry letters (2007-04-13)
Yong Joo Jun, Jung Hee Kim, Su Jin Choi, Hwa Jeong Lee, Moo Jin Jun, Youn Soo Sohn
ABSTRACT
In order to develop a new gene delivery vector, a novel cationic poly(organophosphazene) was synthesized by stepwise nucleophilic substitutions of poly(dichlorophosphazene) with a hydrophilic methoxy-poly(ethylene glycol) (MPEG) as a shielding group and a branched tetra(L-lysine), LysLys(LysEt)(2), as a cationic moiety. The cationic polymer has shown to form a polyplex by DNA condensation and very low in vitro cytotoxicity probably due to the shielding effect of MPEG, which provides a basis for improving the low gene transfection yield of cationic polyphosphazenes.
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)
Sigma-Aldrich
Lys-Lys-Lys, ≥97% (TLC)
Sigma-Aldrich
Lys-Lys-Lys-Lys-Lys, ≥55% peptide basis